Yale Immunofluorescence Protocol
Procedure for Fixation, Immunostaining, and Imaging in 384-well Plates
Download the protocol (PDF)
Go to the Immunofluorescence Tested Antibodies web page
Reagents
Precautions and Disclaimer
Preparation Instructions
Procedure
Results
Reagents
- 384-well view plates (Aurora)
- HUVEC (pooled, Lonza)
- EBM-2 Medium (Lonza)
- Fixation Solution - 16% Paraformaldehyde (PFA) aqueous solution (diluted to 4% in wells)
- 1x PBS w/o Ca2+ and Mg2+ (prepared from powder)
- Donor Calf Serum (DCS)
- Triton® X-100
- Primary antibody
- Secondary Antibody (Alexafluor 647 goat anti-rabbit IgG–highly cross-adsorbed)
- Hoechst 33342
Precautions and Disclaimer
Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Preparation Instructions
Triton X-100 Stock Solution - 10% (v/v) solution in PBS
Antibody Diluent - 10% DCS in PBS
Hoechst 33342 Stock Solution - 10 mg/mL in PBS
Permeabilization and Blocking Solution - 0.2% Triton X-100 and 10% Donor Calf Serum in PBS
Primary Antibody Solutions – Prepare 8 different primary antibody dilutions ranging from 1:50 to 1:6,400 using Antibody Diluent.
Secondary Antibody Solution – dilute secondary antibody 1:1,000 using Antibody Diluent. Add Hoechst 33342 Stock Solution to a final concentration of 1 mg/mL (10,000-fold dilution of Stock Solution).
Procedure
- Cells preparation - HUVEC cells were cultured in EBM-2 medium and plated in 384-well plates at a density of 3,500 cells per well. The plates were then incubated for 24–48 hours until 80–90% confluent before fixation.
- Fixation
- Dilute Fixation Solution (16% PFA) 4-fold in test wells (17 mL of Fixation Solution added to 50 mL of medium in wells).
Note: Alternatively, Fixation Solution can be diluted to 4% PFA with PBS and added directly to wells after aspirating medium. - Incubate for 20 minutes at room temperature.
- Wash once with 1x PBS.
- Store plates at 4 °C until ready to stain.
- Dilute Fixation Solution (16% PFA) 4-fold in test wells (17 mL of Fixation Solution added to 50 mL of medium in wells).
- Immunostaining
- Wash once with 1x PBS and aspirate.
- Add 50 mL of Permeabilization and Blocking Solution to each well.
- Incubate for 20 minutes at room temperature.
- Aspirate Permeabilization and Blocking Solution and add 25 mL of Primary Antibody Solution.
- Incubate for 20 minutes at room temperature.
Note: In some cases, the primary antibody incubation time can be extended to increase signal intensity. - Aspirate Primary Antibody Solution, wash once in 1x PBS, and aspirate.
- Add 25 mL of Secondary Antibody Solution.
- Incubate for 20 minutes at room temperature.
- Wash 4 times with 1x PBS.
- Store plates in 50 mL of 1x PBS at 4 °C until ready to image.
Results
Imaging
All images were acquired using the PerkinElmer® Opera® high-throughput confocal imager. All antibodies were imaged at 8 different dilutions ranging from 1:50 to 1:6400 with 2–3 images taken per well. Plates were imaged at both 20´ (for data acquisition) and 40´ (for high quality image generation). Antibody signal was measured using the 640 nm (red) laser and Hoechst intensity was measured using the 405 nm (violet) laser. In both cases, the exposures were taken separately for each channel to limit fluorescent bleedthrough between the channels. Exposure times varied for each channel with maximums of 2,000 ms for the 640 nm channel and 400 ms for the 405 nm channel.
Data Acquisition
Data was generated using the PerkinElmer Acapella® software package. Intensity values were obtained using the NFkB Translocation algorithm. Nuclear intensity was calculated from the antibody signal in the area overlaid with Hoechst staining. Cytoplasmic intensity was calculated from a cytoplasmic ring region defined by the area outside of the nuclear region. All values represent the average intensity per cell in each field.
Triton is a registered trademark of Dow Chemical Company.
PerkinElmer, Opera, and Acapella are registered trademarks of PerkinElmer Inc.
We warrant that our products conform to the information contained in this and our other publications. Purchaser must determine the suitability of the product(s) for their particular use. Additional terms and conditions may apply. Please see reverse side of the invoice or packing slip.