Merck
LC/MS/MS Analysis of Steroid Hormones in Plasma on Ascentis® Express C18 after Sample Prep using HybridSPE®-Phospholipid
LC/MS/MS Analysis of Steroid Hormones in Plasma on Ascentis® Express C18 after Sample Prep using HybridSPE®-Phospholipid application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
SPE tube or plate
Product No.
Description
Pricing
HybridSPE®-Phospholipid
96-well Plate, bed wt. 50 mg, volume 2 mL, pk of 1
standard
Product No.
Description
Pricing
Aldosterone solution
100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Corticosterone solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
17α-Methyltestosterone solution
1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®
Progesterone solution
1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Testosterone solution
1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
SPE tube/cartridge
HybridSPE®-Phospholipid, 96-well plate, 50 mg/well (575656-U)
sample addition
To each well add 100 μL of plasma followed by 300 μL of precipitation solvent (1% formic acid or 0.5% citric acid in acetonitrile).  (Agitate via vortex for 4 minute, place on vacuum manifold and apply 10" Hg vacuum for 4 minutes. Collect filtrate and analyze directly.)
column
Ascentis® Express C18, 10 cm x 2.1 mm I.D., 2.7 μm particles (53823-U)
gradient
60% B for 3 min, 60% B to 95% B in 5 min, held at 95% B for 2 min
mobile phase
[A] 5 mM ammonium formate pH 4.0 with formic acid; [B] methanol
flow rate
0.3 mL/min
column temp.
50 °C
detector
ESI+, MRM
injection
2 μL
Description
Analysis Note
This application shows the separation of steroid hormones in plasma using LC/MS/MS. HybridSPE-Phospholipid removed endogenous phospholipids and precipitated proteins, thereby allowing rapid, sensitive analysis by LC/MS/MS using an Ascentis Express C18 column. The highest grade LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
HybridSPE is a registered trademark of Merck KGaA, Darmstadt, Germany