HPLC Analysis of Drugs of Abuse on Ascentis® Si (HILIC Mode)
CONDITIONS
column
Ascentis Si, 5 cm x 4.6 mm I.D., 5 μm particles (581511-U)
mobile phase
[A] 0.1% ammonium acetate in water (pH unadjusted): [B] 0.1% ammonium acetate in acetontrile; (10:90, A:B)
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 254 nm
injection
10 μL
sample
as indicated in 0.1% ammonium acetate in 10:90:water:acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany