Merck
  • Crystal structures of the mitochondrial deacylase Sirtuin 4 reveal isoform-specific acyl recognition and regulation features.
Crystal structures of the mitochondrial deacylase Sirtuin 4 reveal isoform-specific acyl recognition and regulation features.
Nature communications (2017-11-16)
Martin Pannek, Zeljko Simic, Matthew Fuszard, Marat Meleshin, Dante Rotili, Antonello Mai, Mike Schutkowski, Clemens Steegborn
ABSTRACT
Sirtuins are evolutionary conserved NAD