- Chemoenzymatic synthesis of 3'-phosphoadenosine-5'-phosphosulfate coupling with an ATP regeneration system.
Chemoenzymatic synthesis of 3'-phosphoadenosine-5'-phosphosulfate coupling with an ATP regeneration system.
Applied microbiology and biotechnology (2017-09-19)
Cuiying An, Long Zhao, Zhaojun Wei, Xianxuan Zhou
PMID28920175
ABSTRACT
3'-Phosphoadenosine-5'-phosphosulfate (PAPS) is the obligate cosubstrate and source of the sulfonate group in the chemoenzymatic synthesis of heparin, a commonly used anticoagulant drug. Previously, using ATP as the substrate, we had developed a one-pot synthesis to prepare PAPS with 47% ATP conversion efficiency. During the reaction, 47% of ATP was converted into the by-product, ADP. Here, to increase the conversion ratio of ATP to PAPS, an ATP regeneration system was developed to couple with PAPS synthesis. In the ATP regeneration system, the chemical compound, monopotassium phosphoenolpyruvate (PEP
MATERIALS