- Preparation of molecularly imprinted polymers for warfarin and coumachlor by multi-step swelling and polymerization method and their imprinting effects.
Preparation of molecularly imprinted polymers for warfarin and coumachlor by multi-step swelling and polymerization method and their imprinting effects.
Journal of chromatography. A (2017-08-19)
Yukari Nakamura, Shizuka Masumoto, Arisa Kubo, Hisami Matsunaga, Jun Haginaka
PMID28818327
ABSTRACT
Monodisperse molecularly imprinted polymers (MIPs) for warfarin (WF) and coumachlor (CC), MIP
MATERIALS