- Synergistic Effect of Pleuromutilins with Other Antimicrobial Agents against
Synergistic Effect of Pleuromutilins with Other Antimicrobial Agents against
Frontiers in pharmacology (2017-09-07)
Chun-Liu Dong, Lin-Xiong Li, Ze-Hua Cui, Shu-Wen Chen, Yan Q Xiong, Jia-Qi Lu, Xiao-Ping Liao, Yuan Gao, Jian Sun, Ya-Hong Liu
PMID28874907
ABSTRACT
Invasive infections due to