Merck
  • Essential role for histone deacetylase 11 (HDAC11) in neutrophil biology.
Essential role for histone deacetylase 11 (HDAC11) in neutrophil biology.
Journal of leukocyte biology (2017-05-28)
Eva Sahakian, Jie Chen, John J Powers, Xianghong Chen, Kamira Maharaj, Susan L Deng, Alex N Achille, Maritza Lienlaf, Hong Wei Wang, Fengdong Cheng, Andressa L Sodré, Allison Distler, Limin Xing, Patricio Perez-Villarroel, Sheng Wei, Alejandro Villagra, Ed Seto, Eduardo M Sotomayor, Pedro Horna, Javier Pinilla-Ibarz
ABSTRACT
Epigenetic changes in chromatin structure have been recently associated with the deregulated expression of critical genes in normal and malignant processes. HDAC11, the newest member of the HDAC family of enzymes, functions as a negative regulator of IL-10 expression in APCs, as previously described by our lab. However, at the present time, its role in other hematopoietic cells, specifically in neutrophils, has not been fully explored. In this report, for the first time, we present a novel physiologic role for HDAC11 as a multifaceted regulator of neutrophils. Thus far, we have been able to demonstrate a lineage-restricted overexpression of HDAC11 in neutrophils and committed neutrophil precursors (promyelocytes). Additionally, we show that HDAC11 appears to associate with the transcription machinery, possibly regulating the expression of inflammatory and migratory genes in neutrophils. Given the prevalence of neutrophils in the peripheral circulation and their central role in the first line of defense, our results highlight a unique and novel role for HDAC11. With the consideration of the emergence of new, selective HDAC11 inhibitors, we believe that our findings will have significant implications in a wide range of diseases spanning malignancies, autoimmunity, and inflammation.
Sigma-Aldrich
Anti-Histone Deacetylase 11 (HDAC11) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Hydrophobin SC3, ≥98% (SDS-GE)