Merck
  • Comparative cytotoxicity of nanoparticles and ions to Escherichia coli in binary mixtures.
Comparative cytotoxicity of nanoparticles and ions to Escherichia coli in binary mixtures.
Journal of environmental sciences (China) (2017-05-10)
Swati Srivastava, Arun Kumar
ABSTRACT
The objective of this study was to understand toxicity of mixture of nanoparticles (NPs) (ZnO and TiO