- Metabolic and Cardiovascular Benefits and Risks of EMD386088-A 5-HT
Metabolic and Cardiovascular Benefits and Risks of EMD386088-A 5-HT
Frontiers in neuroscience (2017-02-24)
Magdalena Kotańska, Joanna Śniecikowska, Magdalena Jastrzębska-Więsek, Marcin Kołaczkowski, Karolina Pytka
PMID28228713
ABSTRACT
Since 5-HT