Merck
  • Fluoranthene degradation and binding mechanism study based on the active-site structure of ring-hydroxylating dioxygenase in Microbacterium paraoxydans JPM1.
Fluoranthene degradation and binding mechanism study based on the active-site structure of ring-hydroxylating dioxygenase in Microbacterium paraoxydans JPM1.
Environmental science and pollution research international (2016-10-11)
Jingnan Jin, Jun Yao, Wenjuan Liu, Qingye Zhang, Jianli Liu
ABSTRACT
In this study, a gram-positive fluoranthene-degrading bacterial strain was isolated from crude oil in Dagang Oilfield and identified as Microbacterium paraoxydans JPM1 by the analysis of 16S rDNA sequence. After 25 days of incubation, the strain JPM1 could degrade 91.78 % of the initial amount of fluoranthene. Moreover, four metabolites 9-fluorenone-1-carboxylic acid, 9-fluorenone, phthalic acid, and benzoic acid were detected in the culture solution. The gene sequence encoding the aromatic-ring-hydroxylating dioxygenase was amplified in the strain JPM1 by PCR. Based on the translated protein sequence, a homology modeling method was applied to build the crystal structure of dioxygenase. Subsequently, the interaction mechanism between fluoranthene and the active site of dioxygenase was simulated and analyzed by molecular docking. Consequently, a feasible degrading pathway of fluoranthene in the strain JPM1 was proposed based on the metabolites and the interaction analyses. Additionally, the thermodynamic analysis showed that the strain JPM1 had high tolerance for fluoranthene, and the influence of fluoranthene for the bacterial growth activity was negligible under 100 to 400 mg L
Sigma-Aldrich
9-Fluorenone, 98%