Merck
  • Synthesis of N-phenyl-N-(3-(piperidin-1-yl)propyl)benzofuran-2-carboxamides as new selective ligands for sigma receptors.
Synthesis of N-phenyl-N-(3-(piperidin-1-yl)propyl)benzofuran-2-carboxamides as new selective ligands for sigma receptors.
Bioorganic & medicinal chemistry (2012-10-23)
Karla-Sue C Marriott, Andrew Z Morrison, Misty Moore, Olarongbe Olubajo, Leonard E Stewart
ABSTRACT
Novel benzofuran-2-carboxamide ligands, which are selective for sigma receptors, have been synthesized via a microwave-assisted Perkin rearrangement reaction and a modified Finkelstein halogen-exchange used to facilitate N-alkylation. The ligands synthesized are the 3-methyl-N-phenyl-N-(3-(piperidin-1-yl)propyl)benzofuran-2-carboxamides (KSCM-1, KSCM-5 and KSCM-11). The benzofuran-2-carboxamide structure was N-arylated and N-alkylated to include both N-phenyl and N-(3-(piperidin-1-yl)propyl substituents, respectively. These new carboxamides exhibit high affinity at the sigma-1 receptor with K(i) values ranging from 7.8 to 34nM. Ligand KSCM-1 with two methoxy substituents at C-5 and C-6 of the benzofuran ring, and K(i)=27.5nM at sigma-1 was found to be more selective for sigma-1 over sigma-2.