Merck
  • Neuroendocrine modulation induced by selective blockade of TNF-alpha in rheumatoid arthritis.
Neuroendocrine modulation induced by selective blockade of TNF-alpha in rheumatoid arthritis.
Annals of the New York Academy of Sciences (2006-07-21)
Gabriele di Comite, Alessandro Marinosci, Paola Di Matteo, Angelo Manfredi, Patrizia Rovere-Querini, Elena Baldissera, Patrizia Aiello, Angelo Corti, Maria Grazia Sabbadini
ABSTRACT
Tumor necrosis factor-alpha (TNFalpha) is a main actor in the pathogenesis of rheumatoid arthritis (RA), interacting with other molecules in complex mechanisms. The neuroendocrine system is known to be involved and Chromogranin A (CHGA) serum levels are elevated in patients with RA. We evaluated the effect of the selective blockade of TNF-alpha, induced by treatment with anti-TNF-alpha monoclonal antibodies (mAbs), on the serum levels of CHGA and on its correlation with TNF-alpha and TNF-alpha receptors (TNFRs) serum levels. Seven patients with RA have been treated with the anti-TNF-alpha mAb, infliximab. We measured the serum levels of TNF-alpha, its receptors (tumor necrosis factor receptor-I [TNFR-I] and tumor necrosis factor receptor-II [TNFR-II]), and CHGA before and during the treatment. We also measured, as a control, the serum levels of CHGA, TNF-alpha, and soluble TNFRs in 14 patients who were being treated with infliximab, adalimumab, or etanercept and in 20 matching negative controls. The serum levels of TNFR-I and TNFR-II, which are a sensitive marker for the TNF-alpha pathway, correlated with those of CHGA before treatment (Pearson's coefficient, respectively, 0.59 and 0.53). Treatment with anti-TNF-alpha mAb provided a significant clinical response in all patients and the correlation between CHGA and TNFR-I and TNFR-II was no more evident during treatment (respectively, -0.09 and -0.07). TNF-alpha blockade allows a clinical effect in patients with RA and modifies the correlation between CHGA and TNFRs, suggesting that TNF-alpha and CHGA reciprocally interfere in the pathogenesis of RA, through intermediate adaptors, whose identification warrants further studies.