Merck
  • Substituted terphenyl compounds as the first class of low molecular weight allosteric inhibitors of the luteinizing hormone receptor.
Substituted terphenyl compounds as the first class of low molecular weight allosteric inhibitors of the luteinizing hormone receptor.
Journal of medicinal chemistry (2009-03-20)
Laura H Heitman, Rajeshwar Narlawar, Henk de Vries, Milou N Willemsen, Dieter Wolfram, Johannes Brussee, Adriaan P Ijzerman
ABSTRACT
The luteinizing hormone (LH) receptor plays an important role in fertility and certain cancers. The endogenous ligands human chorionic gonadotropin (hCG) and LH bind to the large N terminal domain of the receptor. We recently reported on the first radiolabeled low molecular weight (LMW) agonist for this receptor, [(3)H]Org 43553, which was now used to screen for new LMW ligands. We identified a terphenyl derivative that inhibited [(3)H]Org 43553 binding to the receptor, which led us to synthesize a number of derivatives. The most potent compound of this terphenyl series, 24 (LUF5771), was able to increase the dissociation rate of [(3)H]Org 43553 by 3.3-fold (at 10 muM). In a functional assay, the presence of 24 resulted in a 2- to 3-fold lower potency of both Org 43553 and LH. Thus, the compounds presented in this paper are the first LMW ligands that allosterically inhibit the LH receptor.
Sigma-Aldrich
Cyclopentyl isocyanate, 97%