Merck
  • Fluorescent diethylcarbamazine analogues: sites of accumulation in Brugia malayi.
Fluorescent diethylcarbamazine analogues: sites of accumulation in Brugia malayi.
Bioconjugate chemistry (2007-10-12)
Amy Junnila, D Scott Bohle, Roger Prichard, Inna Perepichka, Carla Spina
ABSTRACT
New fluorescein and rhodamine B-labeled antifilarial drug DEC analogues for use in drug localization studies with confocal microscopy have been prepared by a high-yield three-step synthesis. The resulting beta-amine-substituted DEC analogue has a single ethyl substituent which is beta-aminated to accommodate the fluorophore of either fluorescein isothiocyananate or rhodamine B. Confocal microscopy is used to show that the drug accumulates in the adult filarial worms in the pharynx, esophagus, and near the nerve ring of all adults, as well as in the uteri and vulva and the testes of the females and males.
Sigma-Aldrich
4-Methyl-1-piperazinecarbonyl chloride hydrochloride, 97%