Merck
  • Modulation of bone morphogenic protein signaling in T-cells for cancer immunotherapy.
Modulation of bone morphogenic protein signaling in T-cells for cancer immunotherapy.
Journal of immunotoxicology (2013-12-20)
Michal Kuczma, Agnieszka Kurczewska, Piotr Kraj
ABSTRACT
Immunotherapy is becoming an increasingly attractive therapeutic alternative for conventional cancer therapy. In recent years Foxp3(+) regulatory T-cells (T(R)) were identified as the major obstacle to effective cancer immunotherapy. The abundance of these cells in peripheral blood is increased in patients with multiple types of cancer and their prevalence among tumor-infiltrating lymphocytes correlated with poor clinical prognosis. In contrast, removal or inactivation of T(R) cells led to enhanced anti-tumor immune response and better efficacy of cancer vaccines. This study reports that Bone Morphogenic Protein Receptor 1α (BMPR1α, Alk-3) is expressed by activated effector CD4(+) and T(R) cells and modulates functions of both cell types. Bone Morphogenic Proteins (BMPs) belong to the transforming growth factor (TGF)-β family of cytokines that also include TGFβ and activins. BMPs play crucial roles in embryonic development, tissue differentiation and homeostasis, and development of cancer. It was demonstrated that BMPs and activins synergize with TGFβ to regulate thymic T-cell development, maintain T(R) cells, and control peripheral tolerance. Inactivation of BMPR1α in T-cells results in impaired thymic and peripheral generation of T(R) cells. BMPR1α-deficient activated T-cells produced a higher level of interferon (IFN)-γ than BMPR1α-sufficient T-cells. Moreover, transplanted B16 melanoma tumors grew smaller in mice lacking expression of BMPR1α in T-cells and tumors had few infiltrating TR cells and a higher proportion of CD8(+) T-cells than wild-type mice.
Sigma-Aldrich
Anti-IL4 antibody produced in rabbit, affinity isolated antibody