Merck
  • Loss of E-cadherin promotes migration and invasion of cholangiocarcinoma cells and serves as a potential marker of metastasis.
Loss of E-cadherin promotes migration and invasion of cholangiocarcinoma cells and serves as a potential marker of metastasis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-05-29)
Anchalee Techasen, Watcharin Loilome, Nisana Namwat, Narong Khuntikeo, Anucha Puapairoj, Patcharee Jearanaikoon, Hideyuki Saya, Puangrat Yongvanit
ABSTRACT
Tumor progression is characterized by loss of cell adhesion and increase of invasion and metastasis. E-cadherin, a cell adhesion molecule, is frequently downregulated and has been proposed as an important mediator in epithelial-mesenchymal transition (EMT) in tumors. In this study, we investigated the expression of E-cadherin and its association with cancer invasion and prognosis in cholangiocarcinoma (CCA). Immunohistochemistry results demonstrated a statistically significant association between the positive metastasis status with low E-cadherin protein expression in human CCA tissues (P = 0.04). Statistical trends were identified for low E-cadherin level and shorter survival time (P = 0.08). Targeting the E-cadherin expression in CCA cells with siRNA caused upregulation of vimentin, a mesenchymal marker, and disappearance of the E-cadherin/β-catenin adhesion complex from cell membranes. Moreover, migration and invasion abilities of the cells were increased under this condition. These findings suggest that reduction of E-cadherin contributes to CCA progression by attenuating the strength of cellular adhesion, which affects motility as well as regulating the expression of EMT-related genes during CCA invasion and metastasis. Thus, E-cadherin can act as a central modulator of tumor cell phenotype and is a potential metastasis marker in CCA.
Sigma-Aldrich
MISSION® esiRNA, targeting human CDH1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cdh1