Merck
  • A novel MVK missense mutation in one Chinese family with disseminated superficial actinic porokeratosis.
A novel MVK missense mutation in one Chinese family with disseminated superficial actinic porokeratosis.
Molecular biology reports (2014-07-26)
Wen-sheng Lu, Xiao-dong Zheng, Xiu-hua Yao, Lan-fang Zhang, Mu-Qiu Wang, Fa-Xing Jiang, Si-Ping Zhang, Bai Hu
ABSTRACT
Disseminated superficial actinic porokeratosis (DSAP) is a severe chronic autosomal dominant cutaneous disorder with high genetic heterogeneity. mevalonate kinase, (MVK) a gene know to play an important role in regulation of calcium-induced keratinocyte differentiation and proliferation, has recently been suggested as the disease-causing gene for DSAP. Here we report a direct sequencing analysis of this gene in 3 DSAP families, 6 sporadic cases, and 100 unrelated healthy controls. We detected a heterozygous T to A transition at nucleotide 205 in exon 3 of MVK gene in one familial case. This mutation will result in an amino acid change at codon 69 (P.Ser69Thr), which is from a serine codon (TCA) to a threonine codon (ACA). No such mutation was detected in the unaffected family members or the 100 unrelated healthy controls. Our results demonstrated a novel missense mutation in MVK gene. This will be valuable for the diagnosis of DSAP as well as for genetic counseling and prenatal diagnosis of affected families.