Merck
Convenient synthesis of alkyl esters of urocanic acid.
Journal of pharmaceutical sciences (1981-01-01)
K Honda, Y Nishijima
ABSTRACT
Ethyl, n-dodecyl, and n-hexadecyl esters of urocanic acid (4-imidazoleacrylic acid) were prepared from 4-imidazolecarboxaldehyde in satisfactory yields via the Wittig reaction.
Sigma-Aldrich
4-Imidazolecarboxaldehyde, 98%