Merck
  • Cyclodextrin removes cholesterol from mouse sperm and induces capacitation in a protein-free medium.
Cyclodextrin removes cholesterol from mouse sperm and induces capacitation in a protein-free medium.
Biology of reproduction (1998-11-26)
Y H Choi, Y Toyoda
ABSTRACT
Cyclodextrin, which stimulates cholesterol efflux from cells, was examined for its ability to induce capacitation of mouse spermatozoa. A chemically defined, protein-free medium was used for in vitro fertilization of cumulus-free mouse eggs. Fertilization did not occur in modified Krebs-Ringer bicarbonate medium (TYH) supplemented with 1 mg/ml polyvinylalcohol instead of BSA. However, fertilization was observed when spermatozoa were preincubated with methyl-beta-cyclodextrin (MBCD); fertilization rates increased dose-dependently from 0.25 to 0.75 mM MBCD. The fertilization rate decreased when 0.75 mM MBCD was added to both preincubation and fertilization media versus only the preincubation medium (21% vs. 53%); in sharp contrast, fertilization increased when 4 mg/ml BSA was present in both of the media versus the preincubation medium only (66% vs. 25%). At 0.75 mM, 2-hydroxy-beta-cyclodextrin had a lower ability to capacitate spermatozoa in vitro than MBCD (14% vs. 41%). Eggs fertilized by spermatozoa treated with MBCD (0.75 mM) developed to blastocysts (45%, 36 of 80) when cultured in KSOM. When 160 fertilized eggs were transferred to ICR recipients, 62 live offspring were born. After incubation of mouse spermatozoa for 90 min in 0.75 mM MBCD in TYH medium, the cholesterol content of the spermatozoa was significantly (p < 0.01) lower than that of the control (2.27 +/- 0.09 vs. 4.13 +/- 0.09 nmol unesterified cholesterol/10(7) sperm; mean +/- SEM, n = 5). The proportion of capacitated (B pattern) spermatozoa determined by chlortetracycline fluorescence was higher with MBCD treatment for 90 min than for the control (45% vs. 15%; p < 0.01). The proportion of acrosome-reacted (AR pattern) spermatozoa was not different between MBCD treatment and the control. Therefore, MBCD increased capacitation rather than the acrosome reaction of spermatozoa.
Sigma-Aldrich
Chlortetracycline hydrochloride, ≥91.0% dry basis (HPLC)