Merck
  • Identification of a highly potent and selective DNA-dependent protein kinase (DNA-PK) inhibitor (NU7441) by screening of chromenone libraries.
Identification of a highly potent and selective DNA-dependent protein kinase (DNA-PK) inhibitor (NU7441) by screening of chromenone libraries.
Bioorganic & medicinal chemistry letters (2004-11-18)
Justin J J Leahy, Bernard T Golding, Roger J Griffin, Ian R Hardcastle, Caroline Richardson, Laurent Rigoreau, Graeme C M Smith
ABSTRACT
A solution-phase multiple-parallel synthesis approach was employed for the preparation of 6-, 7- and 8-aryl-substituted chromenone libraries, which were screened as inhibitors of the DNA repair enzyme DNA-dependent protein kinase (DNA-PK). These studies resulted in the identification of 8-dibenzothiophen-4-yl-2-morpholin-4-yl-chromen-4-one (NU7441) as a highly potent and selective DNA-PK inhibitor (IC50=14 nM), exhibiting ATP-competitive inhibition kinetics.
Sigma-Aldrich
5′-Bromo-2′-hydroxyacetophenone, 98%
Sigma-Aldrich
NU7441, ≥98% (HPLC)