Merck
  • Diels-Alder reactions of inert aromatic compounds within a self-assembled coordination cage.
Diels-Alder reactions of inert aromatic compounds within a self-assembled coordination cage.
Chemistry, an Asian journal (2011-02-22)
Shinnosuke Horiuchi, Takashi Murase, Makoto Fujita
ABSTRACT
A self-assembled coordination cage serves as a nanometer-sized molecular flask to promote the Diels-Alder reactions of aromatic hydrocarbons with N-cyclohexylmaleimide. The coordination cage accelerated the Diels-Alder reaction of anthracene at the electronically unfavorable, terminal benzene ring to give a compact, cavity-restrained syn-adduct. Activation-parameter measurements for the reactions revealed considerable reduction in the entropy cost, and preorganization of the substrates is a dominant factor in the enhanced reactivity. Owing to this entropy-cost reduction, otherwise-unreactive aromatic compounds, such as naphthalenes or triphenylene, also underwent Diels-Alder reactions in a regio- and stereocontrolled fashion. In the naphthalene Diels-Alder reaction, X-ray crystallographic analysis of the guest-inclusion complex clarified the reinforced orientation and proximity of the substrate pairs before the reaction. A perylene Diels-Alder adduct was stabilized inside the cage and protected from aerial oxidation.
Sigma-Aldrich
N-Cyclohexylmaleimide, 97%