Merck
  • Regioselective synthesis of 4- and 7-alkoxyindoles from 2,3-dihalophenols: application to the preparation of indole inhibitors of phospholipase A2.
Regioselective synthesis of 4- and 7-alkoxyindoles from 2,3-dihalophenols: application to the preparation of indole inhibitors of phospholipase A2.
The Journal of organic chemistry (2007-06-15)
Roberto Sanz, M Pilar Castroviejo, Verónica Guilarte, Antonio Pérez, Francisco J Fañanás
ABSTRACT
An efficient and regioselective synthesis of 4- and 7-alkoxyindoles has been developed from commercially available starting materials such as 3-halophenols and 3-chloroanisole. Directed ortho-metalation followed by two palladium-catalyzed processes, a Sonogashira coupling and a tandem amination/cyclization reaction, allows the synthesis of regiochemically pure 4- and 7-substituted indoles. This strategy has been successfully applied to the preparation of 2-[3-(2-amino-2-oxoacetyl)-1-benzyl-2-ethyl-1H-indol-4-yloxy]acetic acid (LY315920), a known inhibitor of phospholipase A2.
Sigma-Aldrich
3-Chloroanisole, 98%