Merck
Trandolapril/verapamil combination in hypertensive diabetic patients.
Vascular health and risk management (2007-06-23)
José A García Donaire, Luis M Ruilope
ABSTRACT
Cardiovascular diseases are directly affected by arterial hypertension. When associated with diabetes mellitus, the potential deleterious effects are well amplified. Both conditions play a central role in the pathogenesis of coronary artery disease, heart failure, stroke, and renal insufficiency. Prevalence of hypertension is much higher among diabetic than non-diabetic patients, and the hypertensive patient is more likely to develop type 2 diabetes. Current international guidelines recommend aggressive reductions in blood pressure (BP) in hypertensive patients with additional risk factors, including cardiovascular risk factors, and emphasize the relevance of intensive reduction in patients with diabetes mellitus; a goal of 130/80 mm Hg is required. To achieve BP target a combination of antihypertensives will be needed, and the use of long-acting drugs that are able to provide 24-hour efficacy with a once-daily dosing confers the noteworthy advantages of compliance improvement and BP variation lessening. Lower dosages of the individual treatments of the combination therapy can be administered for the same antihypertensive efficiency as that attained with high dosages of monotherapy. Angiotensin-converting enzyme inhibitors and calcium-channel blockers as a combination have theoretically compelling advantages for vessel homeostasis. Trandolapril/verapamil sustained release combination has showed beneficial effects on cardiac and renal systems as well as its antihypertensive efficacy, with no metabolic disturbances. This combination can be considered as an effective therapy for the diabetic hypertensive population.
Sigma-Aldrich
Trandolapril, ≥98% (HPLC), white, powder
Trandolapril, European Pharmacopoeia (EP) Reference Standard