Merck
  • Blockade of endogenous neuraminidase leads to an increase of neuronal excitability and activity-dependent synaptogenesis in the rat hippocampus.
Blockade of endogenous neuraminidase leads to an increase of neuronal excitability and activity-dependent synaptogenesis in the rat hippocampus.
The European journal of neuroscience (2010-11-04)
Elena Isaeva, Irina Lushnikova, Alina Savrasova, Galina Skibo, Gregory L Holmes, Dmytro Isaev
ABSTRACT
Polysialic acids are widely distributed in neuronal tissue. Due to their position on glycoproteins and gangliosides on the outer cell membranes and anionic nature, polysialic acids are involved in multiple cell signaling events. The level of sialylation of the cellular surface is regulated by endogenous neuraminidase (NEU), which catalyses the hydrolysis of terminal sialic acid residues. Using the specific blocker of endogenous NEU, N-acetyl-2,3-dehydro-2-deoxyneuraminic acid (NADNA), we show that downregulation of the endogenous NEU activity causes a significant increase in the level of hippocampal tissue sialylation. Acute application of NADNA increased the firing frequency and amplitude of spontaneous synchronous oscillations, and frequency of multiple unit activity in cultured hippocampal slices. The tonic phase of seizure-like activity in the low-magnesium model of ictogenesis was significantly increased in slices pretreated with NADNA. These data indicate that the degree of synchronization is influenced by the amount of active NEU in cultured hippocampal slices. Pretreatment with NADNA led to an increase of the density of simple and perforated synapses in the hippocampal CA1 stratum radiatum region. Co-incubation of slices with NADNA and high concentrations of calcium eliminated the effect of the NEU blocker on synaptic density, suggesting that synaptogenesis observed following downregulation of the endogenous NEU activity is an activity-dependent process.
Sigma-Aldrich
N-Acetyl-2,3-dehydro-2-deoxyneuraminic acid, ≥93% (TLC)