Merck
  • Importance of C*-H based modes and large amplitude motion effects in vibrational circular dichroism spectra: the case of the chiral adduct of dimethyl fumarate and anthracene.
Importance of C*-H based modes and large amplitude motion effects in vibrational circular dichroism spectra: the case of the chiral adduct of dimethyl fumarate and anthracene.
The journal of physical chemistry. A (2014-05-21)
Marco Passarello, Sergio Abbate, Giovanna Longhi, Susan Lepri, Renzo Ruzziconi, Valentin Paul Nicu
ABSTRACT
The role played by the C*-H based modes (C* being the chiral carbon atom) and the large amplitude motions in the vibrational absorption (VA) and vibrational circular dichroism (VCD) spectra is investigated. The example of an adduct of dimethyl fumarate and anthracene, i.e., dimethyl-(+)-(11R,12R)-9,10-dihydro-9,10-ethanoanthracene-11,12-dicarboxylate, and two deuterated isotopomers thereof specially synthesized for this goal, are considered. By comparing the experimental and DFT calculated spectra of the undeuterated and deuterated species, we demonstrate that the C*-H bending, rocking, and stretching modes in the VA and VCD spectra are clearly identified in well defined spectroscopic features. Further, significant information about the conformer distribution is gathered by analyzing the VA and VCD data of both the fingerprint and the C-H stretching regions, with particular attention paid to the band shape data. Effects related to the large amplitude motions of the two methoxy moieties have been simulated by performing linear transit (LT) calculations, which consists of varying systematically the relative positions of the two methoxy moieties and calculating VCD spectra for the partially optimized structures obtained in this way. The LT method allows one to improve the quality of calculated spectra, as compared to experimental results, especially in regard to relative intensities and bandwidths.
Sigma-Aldrich
Dimethyl fumarate, 97%