Merck
  • Synthesis of α-tribromomethylamines via Mg-mediated addition of bromoform to imines.
Synthesis of α-tribromomethylamines via Mg-mediated addition of bromoform to imines.
Organic & biomolecular chemistry (2014-03-29)
Elumalai Gopi, Irishi N N Namboothiri
ABSTRACT
Mg-mediated addition of bromoform to electron deficient imines such as N-sulfonylimines affords α-tribromomethylated N-sulfonylamines in good to excellent yields. The procedure could be further simplified by transforming the imine precursors, α-sulfonyl-N-tosyl- and Boc-amines, in one pot to the corresponding α-tribromomethyl derivatives. Facile removal of the Boc protecting group in nearly quantitative yield and a silver carbonate mediated monodebromination have also been demonstrated.
Sigma-Aldrich
Bromoform, contains 60-120 ppm 2-methyl-2-butene as stabilizer, 99%