Merck
  • [Analytical application of phosphotungstic acid complexes. Synthesis and characterisation].
[Analytical application of phosphotungstic acid complexes. Synthesis and characterisation].
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi (2007-11-07)
M Apostu, V Dorneanu, Nela Bibire
ABSTRACT
Phosphotungstic acid forms salt type ionic association complexes with a large number of medicinal drugs that contain cation forming proton acceptor moieties. Ranitidine, nizatidine and famotidine complexes with phosphotungstic acid were synthesized and subsequently characterized by their melting point, water solubility, specific absorbance, UV spectra and IR spectra. These sparingly soluble complexes were applied for the fabrication of membrane selective electrodes for quantitative potentiometric determination of ranitidine, nizatidine and famotidine. There have been synthesized phosphotungstic acid complexes with uses in fabrication of membrane selective electrodes for quantitative potentiometric determination of ranitidine, nizatidine and famotidine.