Merck
  • Novel trisaccharide fatty acid ester identified from the fruits of Morinda citrifolia (Noni).
Novel trisaccharide fatty acid ester identified from the fruits of Morinda citrifolia (Noni).
Journal of agricultural and food chemistry (1999-12-22)
M Wang, H Kikuzaki, K Csiszar, C D Boyd, A Maunakea, S F Fong, G Ghai, R T Rosen, N Nakatani, C T Ho
ABSTRACT
Two known glycosides and a novel trisaccharide fatty acid ester were isolated from the n-butanol-soluble fraction of the fruits of Morinda citrifolia (noni). Structure determination was carried out by spectral techniques such as MS, IR, NMR, and 2D-NMR. The novel trisaccharide fatty acid ester was elucidated as 2, 6-di-O-(beta-D-glucopyranosyl)-1-O-octanoyl-beta-D-glucopyranose. The known compounds were identified as rutin and asperulosidic acid.
Sigma-Aldrich
Asperuloside, ≥95% (LC/MS-ELSD)