Merck
  • Stopped-flow kinetic study of the aroxyl radical-scavenging action of catechins and vitamin C in ethanol and micellar solutions.
Stopped-flow kinetic study of the aroxyl radical-scavenging action of catechins and vitamin C in ethanol and micellar solutions.
Journal of agricultural and food chemistry (2008-05-27)
Shuji Mitani, Aya Ouchi, Emi Watanabe, Yu Kanesaki, Shin-Ichi Nagaoka, Kazuo Mukai
ABSTRACT
Kinetic study of the aroxyl radical-scavenging action of catechins (epicatechin (EC), epicatechin gallate (ECG), epigallocatechin (EGC), and epigallocatechin gallate (EGCG)) and related compounds (methyl gallate (MG), 4-methylcatechol (MC), and 5-methoxyresorcinol (MR)) has been performed. The second-order rate constant ( k s) for the reaction of these antioxidants with aroxyl radical has been measured in ethanol and aqueous Triton X-100 micellar solution (5.0 wt %). The k s values decreased in the order of EGCG > EGC > MC > ECG > EC > MG > MR in ethanol, indicating that the reactivity of the OH groups in catechins decreased in the order of pyrogallol B-ring > catechol B-ring > gallate G-ring > resorcinol A-ring. The structure-activity relationship in the free radical-scavenging reaction by catechins has been clarified by the detailed analyses of the pH dependence of k s values. From the results, the p K a values have been determined for catechins. The monoanion form at catechol B- and resorcinol A-rings and dianion form at pyrogallol B- and gallate G-rings show the highest activity for free radical scavenging. It was found that the free radical-scavenging activities of catechins are 3.2-13 times larger than that of vitamin C at pH 7.0.
Sigma-Aldrich
5-Methoxyresorcinol, 98%