Merck
Expedient parallel synthesis of 2-amino-4-heteroarylpyrimidines.
Organic letters (2005-09-09)
Matthew G Bursavich, Sabrina Lombardi, Adam M Gilbert
ABSTRACT
[reaction: see text] An expedient synthesis of diverse 2-amino-4-heteroarylpyrimidines via a 2-chloropyrimidine intermediate is described. A series of potentially biologically active analogues have been synthesized in two parallel steps to afford focused arrays.
Sigma-Aldrich
2-Chloropyrimidine, 95%