Merck
  • Novel 2,2-dioxide-4,4-disubstituted-1,3-H-2,1,3-benzothiadiazines as non-nucleoside reverse transcriptase inhibitors.
Novel 2,2-dioxide-4,4-disubstituted-1,3-H-2,1,3-benzothiadiazines as non-nucleoside reverse transcriptase inhibitors.
Bioorganic & medicinal chemistry letters (2000-02-15)
J W Corbett, L A Gearhart, S S Ko, J D Rodgers, B C Cordova, R M Klabe, S K Erickson-Viitanen
ABSTRACT
Benzothiadiazine non-nucleoside reverse transcriptase inhibitors (NNRTIs) of HIV have been synthesized via a novel process to afford active inhibitors, with the most potent compound exhibiting an IC90 = 180 nM in a whole cell assay. The 2,2-dioxide-1H-2,1,3-benzothiadiazine ring system was constructed in one step from 2-amino-5-chlorobenzonitrile.
Sigma-Aldrich
2-Amino-5-chlorobenzonitrile, 98%