Merck
Synthesis and antimicrobial activity of new adamantane derivatives II.
Farmaco (Societa chimica italiana : 1989) (2001-01-11)
A Orzeszko, B Kamińska, G Orzeszko, B J Starościak
ABSTRACT
A series of new derivatives of adamantane was synthesised. The new compound 4-(adamant-1-ylethylenoxycarbonyl)phthalanhydride obtained from 1-adamantaneethanol and trimellitic anhydride chloride, as well as 4-(adamant-1-ylmethylenoxycarbonyl)phthalanhydride, appeared very useful for preparation of a number of N-substituted phthalimides. Antimicrobial activity of newly obtained derivatives such as, for example, 4-(adamant-1-ylethylenoxycarbonyl)-N-(L-phenylalanyl)phthalimide or 4-(adamant-1-ylmethylenoxycarbonyl)-N-(L-leucyl)-phthalimide was tested against Staphylococcus aureus, Bacillus sp., Micrococcus flavus and Enterococcus faecium. The minimal inhibitory concentrations for these compounds against Bacillus cereus were 15 and 8 microg/ml, respectively.