Merck
  • The disposition and metabolism of a hypnotic benzodiazepine, quazepam, in the hamster and mouse.
The disposition and metabolism of a hypnotic benzodiazepine, quazepam, in the hamster and mouse.
Drug metabolism and disposition: the biological fate of chemicals (1984-07-01)
J Hilbert, B Pramanik, S Symchowicz, N Zampaglione
ABSTRACT
The disposition of 14C-quazepam (7-chloro-(2,2,2-trifluoroethyl) [5-14C]-5-o-fluorophenyl-1,3-dihydro-2H-1,4-benzodiazepin-2-thione), a new benzodiazepine hypnotic, was studied in hamsters and mice after iv and po dosing. In both species, quazepam was rapidly absorbed, as indicated by the plasma Cmax being reached within 1 hr of an oral dose (5 mg/kg). Also, radioactivity is essentially completely absorbed in both species, since the percentage of dose excreted in the urine was not dependent on the route of drug administration. Radioactivity was widely distributed in the tissues of both species; however, it was concentrated (relative to plasma) only in the liver and kidneys. In hamsters, 66-77% of the radioactivity was excreted within 48 hr, and 97% within 7 days of dosing (57% found in urine and 40% in feces after iv; 54% in urine and 43% in feces after po dosing). In mice, 86-88% of the radioactivity was excreted within 24 hr, and 98% within 4 days of dosing (43% in urine and 56% in feces after iv, 37% in urine and 61% in feces after po dosing). In both species, plasma levels of quazepam, measured by GLC, accounted for a very small percentage of plasma radioactivity and the elimination half-life was short (2.4 hr in hamster and 1.2 hr in mice), indicating extensive first pass metabolism for this drug. TLC analysis of plasma and urine extracts from both species showed biotransformation of quazepam involved substitution of oxygen for sulfur, followed by: (a) N-dealkylation, 3-hydroxylation, and conjugation or (b) 3-hydroxylation and conjugation.(ABSTRACT TRUNCATED AT 250 WORDS)
Product Description
Supelco
Desalkylflurazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®