Merck
  • [Pharmacological features of naltrexone and its use in the treatment of alcohol dependence].
[Pharmacological features of naltrexone and its use in the treatment of alcohol dependence].
Psychiatria polska (2013-07-31)
Przemysław Bieńkowski
ABSTRACT
Clinical studies performed over the last decades have indicated that combined therapy, involving pharmaco- and psychotherapy, is an optimal approach to alcohol dependence. Both pharmaco- and psychotherapy should be personalised with a careful balance between patient's needs and his/her clinical characteristics. The aim of the present article is to review the basic pharmacological features of naltrexone and its use in the treatment of alcohol dependence.
Product Description
Supelco
Naltrexone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®