Merck
  • Microquantification of cholesterol and cholesteryl esters in rat peritoneal macrophages by reverse-phase high-performance liquid chromatography.
Microquantification of cholesterol and cholesteryl esters in rat peritoneal macrophages by reverse-phase high-performance liquid chromatography.
Analytical biochemistry (1990-03-01)
N Araki, S Horiuchi, A T Rahim, K Takata, Y Morino
ABSTRACT
A simple and rapid method for the microquantification of cholesterol and cholesteryl esters by reverse-phase high performance liquid chromatography has been established. Comparison of elution patterns of authentic cholesterol and cholesteryl esters revealed that a mu Bondasphere reverse-phase C8 (300-A) column was more suitable than a corresponding reverse-phase C4 or C18 column in terms of rapidity and sensitivity. Recovery of cholesterol and cholesteryl esters from a C8 column was greater than 98% when determined either by radioactive cholesterol and cholesteryl oleate or by cholesteryl heptadecanoate. The sensitivity of the quantification ranged from 5 ng to 50 micrograms for both cholesterol and cholesteryl esters. This method was applied to determination of cellular cholesterol and cholesteryl esters of rat peritoneal macrophages. Lipid extracts of these cells were found to contain 38.01 +/- 2.60 micrograms of cholesterol and 3.18 +/- 0.36 micrograms of cholesteryl esters per milligram of cell protein. When the cells were loaded with cholesteryl esters by incubation for 24 h with various concentrations of acetylated low-density lipoprotein, a cellular level of cholesteryl esters showed a dose-dependent increase and reached a maximal level of 106.60 +/- 3.05 micrograms/mg cell protein. Thus, the present method is useful for the microquantification of cholesterol and cholesteryl esters from lipid extracts of biological samples.
Sigma-Aldrich
Cholesteryl arachidonate, ≥95% (HPLC; detection at 205 nm), viscous liquid