MilliporeSigma
  • Protein crystal structures with ferrocene and ruthenocene-based enzyme inhibitors.
Protein crystal structures with ferrocene and ruthenocene-based enzyme inhibitors.
Chemical communications (Cambridge, England) (2012-01-20)
Adam J Salmon, Michael L Williams, Andreas Hofmann, Sally-Ann Poulsen
ABSTRACT
We have determined the protein X-ray crystal structures of four organometallic inhibitors in complex with their target enzyme carbonic anhydrase II. The barrel-shaped hydrophobic ferrocene and ruthenocene moieties have provided a structure-based avenue to better occupy the hydrophobic binding patch within the enzyme active site.
Sigma-Aldrich
Bis(cyclopentadienyl)ruthenium(II), 97%