- A rapid and efficient ultrasound-assisted synthesis of 5,5-diphenylhydantoins and 5,5-diphenyl-2-thiohydantoins.
A rapid and efficient ultrasound-assisted synthesis of 5,5-diphenylhydantoins and 5,5-diphenyl-2-thiohydantoins.
Ultrasonics sonochemistry (2010-10-06)
Naimeh Moshtael Arani, Javad Safari
PMID20920873
ABSTRACT
To obtain a rapid, efficient and mild synthesis of 5,5-diphenylhydantoin and 5,5-diphenyl-2-thiohydantoin derivatives, ultrasonic irradiation has been applied to the reaction mixtures containing substituted benzils and urea or thiourea derivatives catalyzed by KOH in DMSO/H(2)O, which allowed us to achieve products at room temperature in a good yield and short time without any side product. This convenient procedure will allow a further increase of the diversity within the hydantoin family.