- Ligand-to-metal ratio controlled assembly of nanoporous metal-organic frameworks.
Ligand-to-metal ratio controlled assembly of nanoporous metal-organic frameworks.
Chemical communications (Cambridge, England) (2008-06-07)
Jian-Guo Lin, Yan-Yan Xu, Ling Qiu, Shuang-Quan Zang, Chang-Sheng Lu, Chun-Ying Duan, Yi-Zhi Li, Song Gao, Qing-Jin Meng
PMID18535699
ABSTRACT
Two bilayered metal-organic frameworks with nanoporous channels were synthesized at different ligand-to-metal ratios, which demonstrated an interesting crystal-to-crystal transformation property and a special fluorescent response to the different guest molecules included.