Merck
  • A new sigma ligand, (+/-)-PPCC, antagonizes kappa opioid receptor-mediated antinociceptive effect.
A new sigma ligand, (+/-)-PPCC, antagonizes kappa opioid receptor-mediated antinociceptive effect.
Life sciences (2008-02-12)
Orazio Prezzavento, Carmela Parenti, Agostino Marrazzo, Simone Ronsisvalle, Franco Vittorio, Giuseppina Aricò, Giovanna M Scoto, Giuseppe Ronsisvalle
ABSTRACT
The compound (1R,2S/1S,2R)-2-[4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl) cyclopropanecarboxylate [(+/-)-PPCC] is a ligand with high affinity for sigma (sigma) sites of which the selectivity towards several other receptor systems has been demonstrated. Given the existence of a relationship between the sigma system and the kappa opioid (KOP)-mediated analgesia, to characterize the pharmacological properties of (+/-)-PPCC we analyzed its influence on the analgesic effect of the systemic injected kappa agonist (-)-U-50,488H comparing the effects with those shown by (+)-pentazocine and BD1047. The results demonstrate that the systemic administration of (+/-)-PPCC (1 mg/kg s.c.) does not modify basal tail-flick latency. Pre-treatment with (+/-)-PPCC, at the same dose, significantly decreased the antinociceptive effect of (-)-U-50,488H, analogously to the sigma compounds used. This study confirms that (+/-)-PPCC plays the role of sigma agonist in this model and strengthens the hypothesis of the sigma receptor modulatory role on KOP-mediated analgesia.
Sigma-Aldrich
Cyclopropanecarboxylic acid, 95%