Merck
  • Chemical composition and in vitro evaluation of antimicrobial and anti-acetylcholinesterase properties of the flower oil of Ferula lutea.
Chemical composition and in vitro evaluation of antimicrobial and anti-acetylcholinesterase properties of the flower oil of Ferula lutea.
Natural product communications (2012-08-23)
Mansour Znati, Aymen Jabrane, Hafedh Hajlaoui, Fethia Harzallah-Skhiri, Jalloul Bouajila, Joseph Casanova, Hichem Ben Jannet
ABSTRACT
The present work describes the chemical composition and evaluates the antimicrobial and the anti-acetylcholinesterase properties of the flower oil from the Tunisian Ferula lutea obtained by hydrodistillation and analyzed by combination of GC/FID and GC/MS. The chemical composition of the flower oil of this species is reported for the first time. Seventeen compounds were identified accounting for 94.3% of the total oil. The chemical composition of this essential oil was characterized by a high proportion of monoterpene hydrocarbons (80.4%) among which delta-3-carene (31.2%) and alpha-pinene (25.8%) were the predominant compounds. The oxygenated monoterpenes represent the second major fraction (12.0%), 2,3,6-trimethylbenzaldehyde (10.9%) being the predominant one. Furthermore, the isolated oil was tested for its antimicrobial activity using the disc-diffusion and the microdilution assays against six Gram-positive and five Gram-negative bacteria as well as towards eight Candida species. It was found that flower oil of F. lutea exhibited interesting antibacterial and anticandidal activity (MIC = 39 mcirog/mL against Escherichia coli, Staphylococcus aureus and S. epidermidis and MIC = 156 microg/mL against Candida albicans). The anti-acetylcholinesterase effect of this oil was also evaluated in this work. Results showed that this oil exhibits significant activity (IC50 =70.25 +/- 5.41 microg/mL).
Product Description
α-Pinene, primary reference standard
Sigma-Aldrich
α-Pinene, 98%