Merck
8-Aminocaprylic acid.
Acta crystallographica. Section C, Crystal structure communications (1998-09-18)
A J Dobson, R E Gerkin
ABSTRACT
The title acid, 8-aminooctanoic acid, C8H17NO2, crystallized in the centrosymmetric space group P2(1)/n in the zwitterionic form. The three H atoms involved in hydrogen bonding are ordered. The five intermolecular N-H...O hydrogen bonds have N...O distances ranging from 2.752 (2) to 3.258 (2) A and N-H...O angles ranging from 131 (2) to 165 (2) degrees. Each molecule is linked to six neighboring molecules by a total of ten hydrogen bonds. A complex network of hydrogen bonds ensues in which chains predominate.
Sigma-Aldrich
8-Aminooctanoic acid, 99%