Merck
  • Na+,K+-ATPase inhibitors in rat urine: origins and physiological significance.
Na+,K+-ATPase inhibitors in rat urine: origins and physiological significance.
Clinical and experimental hypertension (New York, N.Y. : 1993) (1998-07-31)
M Tamura, E J Landon, T Inagami
ABSTRACT
To identify the origins and structures of mammalian tissue-derived Na+,K+-ATPase inhibitors, we investigated the tissue distribution of inhibitors in rats. Among many tissues tested, urine was found to contain high levels of many inhibitors. The structures of the two major inhibitors were identified as neoconvalloside and periplogenin monorhamnoside, which are derivatives of strophanthidin. Urinary levels of these inhibitors, however, decreased considerably after changing the diet from the regular diet to purified synthetic diet, suggesting that the majority of the urinary inhibitors are of dietary origin. Investigation of the ingredients of the diet further revealed that alfalfa meal and ground oats are the major sources of these cardiac glycosides. As to the physiological relevance of the cardiac glycosides, a low concentration (1-50 nM) of ouabain dose-dependently enhanced aldosterone secretion from adrenal glomerulosa cells by an increase in local renin release. Ouabain was also found to be involved in AT2 receptor-specific expression in rat PC12W cells through an increment in intracellular Na+. These results suggest that Na+,K+-ATPase inhibitors, regardless of the source, are involved in the regulation of blood pressure.
Sigma-Aldrich
Digitoxigenin