Merck
Developmental toxicity of dibasic esters by inhalation in the rat.
Drug and chemical toxicology (1995-11-01)
L Alvarez, C Driscoll, D P Kelly, R E Staples, N C Chromey, G L Kennedy
ABSTRACT
Dibasic esters (DBE) are a mixture of 3 carboxylic acid esters which are used in the paint and coatings industry. In this study, groups of pregnant Crl:CD BR rats were exposed to either 0.16, 0.4, or 1.0 mg DBE/L by inhalation for 6 hr/day from Days 7 through 16 of gestation (day in which copulation plug was detected was designated Day 1G). A control group of chambered pregnant rats was exposed simultaneously to air only. All female rats were euthanized on Day 21G and the fetuses were examined. A suppression of both food consumption and the rate of body weight gain was seen in the 0.4 and 1.0 mg/L groups during the first 6 exposure days. Staining on the fur and perineal area was seen in rats exposed to 1.0 mg/L and liver weight decreases, although not statistically significant, occurred in the 2 high exposure groups. None of the reproductive parameters were altered in any of the groups and no fetal effects were detected. DBE is not a developmental toxin in the rat following inhalation exposures as high as 1.0 mg DBE/L during the period of organogenesis.