Merck
  • Free-radical damage to lipids, amino acids, carbohydrates and nucleic acids determined by thiobarbituric acid reactivity.
Free-radical damage to lipids, amino acids, carbohydrates and nucleic acids determined by thiobarbituric acid reactivity.
The International journal of biochemistry (1982-01-01)
J M Gutteridge
ABSTRACT
1. The thiobarbituric acid (TBA) reaction, widely applied to the detection of autoxidation in polyunsaturated fatty acids, can be used to measure free-radical damage to amino acids, carbohydrates and nucleic acids. 2. In all of these systems malondialdehyde (MDA) is predominately formed from intermediate precursor molecules which break down during the acid-heating stage of the TBA test. 3. The acid reagent used to bring about these decompositions appears to be critical.
Sigma-Aldrich
2-Methylindole, 98%