Merck
Liquid chromatographic determination of barbaloin (aloin) in foods.
Journal - Association of Official Analytical Chemists (1985-05-01)
M Yamamoto, M Ishikawa, T Masui, H Nakazawa, Y Kabasawa
ABSTRACT
A simple and rapid liquid chromatographic method is described for the determination of barbaloin (aloin, 10-D-glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-9(10H)-anthraceno ne) in foods. Barbaloin is extracted with water from foods containing aloe and the extract is cleaned up on a disposable cartridge by using methanol-water (55 + 45) as eluant. The eluted barbaloin is separated by liquid chromatography on a YMC A-302 column with methanol-water (50 + 50) mobile phase, and detected at 293 nm. Recoveries of barbaloin added to foods at the levels of 0.05 and 0.50 mg/g were 94.4-100%. Assay results for commercial food samples indicated that the present method is applicable to a variety of foods supplemented with aloe.
Sigma-Aldrich
Aloin, from Curacao aloe, ~50%
Sigma-Aldrich
Aloin, from Aloe barbadensis Miller leaves, ≥97%