Merck
  • A Pan-BCL2 inhibitor renders bone-marrow-resident human leukemia stem cells sensitive to tyrosine kinase inhibition.
A Pan-BCL2 inhibitor renders bone-marrow-resident human leukemia stem cells sensitive to tyrosine kinase inhibition.
Cell stem cell (2013-01-22)
Daniel J Goff, Angela Court Recart, Anil Sadarangani, Hye-Jung Chun, Christian L Barrett, Maryla Krajewska, Heather Leu, Janine Low-Marchelli, Wenxue Ma, Alice Y Shih, Jun Wei, Dayong Zhai, Ifat Geron, Minya Pu, Lei Bao, Ryan Chuang, Larisa Balaian, Jason Gotlib, Mark Minden, Giovanni Martinelli, Jessica Rusert, Kim-Hien Dao, Kamran Shazand, Peggy Wentworth, Kristen M Smith, Christina A M Jamieson, Sheldon R Morris, Karen Messer, Lawrence S B Goldstein, Thomas J Hudson, Marco Marra, Kelly A Frazer, Maurizio Pellecchia, John C Reed, Catriona H M Jamieson
ABSTRACT
Leukemia stem cells (LSCs) play a pivotal role in the resistance of chronic myeloid leukemia (CML) to tyrosine kinase inhibitors (TKIs) and its progression to blast crisis (BC), in part, through the alternative splicing of self-renewal and survival genes. To elucidate splice-isoform regulators of human BC LSC maintenance, we performed whole-transcriptome RNA sequencing, splice-isoform-specific quantitative RT-PCR (qRT-PCR), nanoproteomics, stromal coculture, and BC LSC xenotransplantation analyses. Cumulatively, these studies show that the alternative splicing of multiple prosurvival BCL2 family genes promotes malignant transformation of myeloid progenitors into BC LSCS that are quiescent in the marrow niche and that contribute to therapeutic resistance. Notably, sabutoclax, a pan-BCL2 inhibitor, renders marrow-niche-resident BC LSCs sensitive to TKIs at doses that spare normal progenitors. These findings underscore the importance of alternative BCL2 family splice-isoform expression in BC LSC maintenance and suggest that the combinatorial inhibition of prosurvival BCL2 family proteins and BCR-ABL may eliminate dormant LSCs and obviate resistance.
Sigma-Aldrich
(±)-Gossypol from cotton seeds, ≥95% (HPLC)