Merck
  • Geniposide, from Gardenia jasminoides Ellis, inhibits the inflammatory response in the primary mouse macrophages and mouse models.
Geniposide, from Gardenia jasminoides Ellis, inhibits the inflammatory response in the primary mouse macrophages and mouse models.
International immunopharmacology (2012-08-11)
Yunhe Fu, Bo Liu, Jinhua Liu, Zhicheng Liu, Dejie Liang, Fengyang Li, Depeng Li, Yongguo Cao, Xichen Zhang, Naisheng Zhang, Zhengtao Yang
ABSTRACT
Geniposide, a main iridoid glucoside component of gardenia fruit, has been known to exhibit antibacterial, anti-inflammatory and other important therapeutic activities. The objective of this study was to investigate the protective effects of geniposide on inflammation in lipopolysaccharide (LPS) stimulated primary mouse macrophages in vitro and LPS induced lung injury model in vivo. The expression of pro-inflammatory cytokines was determined by enzyme-linked immunosorbent assay (ELISA). Nuclear factor-kappa B (NF-κB), inhibitory kappa B (IκBα) protein, p38, extracellular signal-regulated kinase (ERK), c-Jun N-terminal kinase (JNK) and Toll-like receptor 4 (TLR4) were determined by Western blot. Further analysis was carried out in mTLR4 and mMD-2 co-transfected HEK293 cells. The results showed that geniposide markedly inhibited the LPS-induced TNF-α, IL-6 and IL-1β production both in vitro and in vivo. Geniposide blocked the phosphorylation of IκBα, p65, p38, ERK and JNK in LPS stimulated primary mouse macrophages. Furthermore, geniposide inhibited the expression of TLR4 in LPS stimulated primary mouse macrophages and inhibited the LPS-induced IL-8 production in HEK293-mTLR4/MD-2 cells. In vivo study, it was also observed that geniposide attenuated lung histopathologic changes in the mouse models. These results suggest that geniposide exerts an anti-inflammatory property by down-regulating the expression of TLR4 up-regulated by LPS. Geniposide is highly effective in inhibiting acute lung injury and may be a promising potential therapeutic reagent for acute lung injury treatment.
Sigma-Aldrich
Geniposide, ≥98% (HPLC)