Merck
  • Astragaloside IV, a novel antioxidant, prevents glucose-induced podocyte apoptosis in vitro and in vivo.
Astragaloside IV, a novel antioxidant, prevents glucose-induced podocyte apoptosis in vitro and in vivo.
PloS one (2012-06-30)
Dingkun Gui, Yongping Guo, Feng Wang, Wei Liu, Jianguo Chen, Yifang Chen, Jianhua Huang, Niansong Wang
ABSTRACT
Glucose-induced reactive oxygen species (ROS) production initiates podocyte apoptosis, which represents a novel early mechanism leading to diabetic nephropathy (DN). Here, we tested the hypothesis that Astragaloside IV(AS-IV) exerts antioxidant and antiapoptotic effects on podocytes under diabetic conditions. Apoptosis, albuminuria, ROS generation, caspase-3 activity and cleavage, as well as Bax and Bcl-2 mRNA and protein expression were measured in vitro and in vivo. Cultured podocytes were exposed to high glucose (HG) with 50, 100 and 200 µg/ml of AS-IV for 24 h. AS-IV significantly attenuated HG-induced podocyte apoptosis and ROS production. This antiapoptotic effect was associated with restoration of Bax and Bcl-2 expression, as well as inhibition of caspase-3 activation and overexpression. In streptozotocin (STZ)-induced diabetic rats, severe hyperglycemia and albuminuria were developed. Increased apoptosis, Bax expression, caspase-3 activity and cleavage while decreased Bcl-2 expression were detected in diabetic rats. However, pretreatment with AS-IV (2.5, 5, 10 mg·kg(-1)·d(-1)) for 14 weeks ameliorated podocyte apoptosis, caspase-3 activation, renal histopathology, podocyte foot process effacement, albuminuria and oxidative stress. Expression of Bax and Bcl-2 mRNA and protein in kidney cortex was partially restored by AS-IV pretreatment. These findings suggested AS-IV, a novel antioxidant, to prevent Glucose-Induced podocyte apoptosis partly through restoring the balance of Bax and Bcl-2 expression and inhibiting caspase-3 activation.
Product Description
Millipore
Astragaloside IV, >98.0%