Merck
  • Dynamics in self-assembled organic monolayers at the liquid/solid interface revealed by scanning tunneling microscopy.
Dynamics in self-assembled organic monolayers at the liquid/solid interface revealed by scanning tunneling microscopy.
Chimia (2012-05-02)
Steven De Feyter, Hong Xu, Kunal Mali
ABSTRACT
The liquid/solid interface provides an interesting medium for molecular self-assembly and scanning tunneling microscopy is the preferred technique to analyse the structural features of the surface-supported self-assembled monolayers in this medium. An interesting aspect is the phenomenon of molecular dynamics at the liquid/solid interface. In this mini-review, we report on our efforts and strategies to investigate and even induce molecular dynamics at the liquid/solid interface, bringing insight to various kinds of processes such as conformational, translational and adsorption/desorption dynamics.
Sigma-Aldrich
Isophthalic acid, 99%